3-chloro-4-hydroxypyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Chloropyridin-4-ol

072071-1G 072071-5G
102522-942EA 83.08 USD
102522-942 102522-944
3-chloro-4-hydroxypyridine ≥95%
3-chloro-4-hydroxypyridine

Hazard: Irritant

Formula: C₅H₄ClNO
MW: 129.543 g/mol
MDL Number: MFCD04114248
CAS Number: 89284-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR