2-Fluoro-5-hydroxypyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-5-pyridinol, 6-Fluoropyridin-3-ol

072069-1G 072069-5G
102522-928EA 20 USD
102522-928 102522-930
2-Fluoro-5-hydroxypyridine ≥95%
2-Fluoro-5-hydroxypyridine

Hazard: Irritant

Formula: C₅H₄FNO
Melting Pt: 150-153°C
MDL Number: MFCD01075040
CAS Number: 55758-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR