tert-Butyl methyl(piperidin-4-yl)carbamate ≥95%

Supplier: Matrix Scientific

072068-1G 072068-5G 072068-10G
102522-922EA 284.62 USD
102522-922 102522-924 102522-926
tert-Butyl methyl(piperidin-4-yl)carbamate ≥95%
tert-Butyl methyl(piperidin-4-yl)carbamate

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD02683051
CAS Number: 108612-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR