4,6-Dichloroisatin ≥95%

Supplier: Matrix Scientific

Synonyms: CFL-120

072066-1G 072066-5G
102522-912EA 52.31 USD
102522-912 102522-914
4,6-Dichloroisatin ≥95%
4,6-Dichloroisatin

Hazard: Irritant

Formula: C₈H₃Cl₂NO₂
MW: 216.0200 g/mol
MDL Number: MFCD01971146
CAS Number: 18711-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR