6-Chlorosalicylaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-hydroxybenzaldehyde

072065-1G 072065-5G
102522-906EA 255.38 USD
102522-906 102522-908
6-Chlorosalicylaldehyde ≥95%
6-Chlorosalicylaldehyde

Hazard: Irritant

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
MDL Number: MFCD01646166
CAS Number: 18362-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR