6-Chloro-2(1H)-quinoxalinone ≥95%

Supplier: Matrix Scientific

072064-1G 072064-5G
102522-902EA 640 USD
102522-902 102522-904
6-Chloro-2(1H)-quinoxalinone ≥95%
6-Chloro-2(1H)-quinoxalinone

Hazard: Irritant

Formula: C₈H₅ClN₂O
MW: 180.59 g/mol
MDL Number: MFCD06200849
CAS Number: 2427-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR