2-(2,4-Dichlorophenyl)thiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

072061-1G 072061-5G
102522-888EA 710.77 USD
102522-888 102522-890
2-(2,4-Dichlorophenyl)thiazole-4-carboxylic acid ≥95%
2-(2,4-Dichlorophenyl)thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₅Cl₂NO₂S
MDL Number: MFCD07376332
CAS Number: 1094355-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR