Ethyl-2-(2,4-dichlorophenyl)thiazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
072060-1G
072060-5G
102522-884EA
526.15
USD
102522-884
102522-886
Ethyl-2-(2,4-dichlorophenyl)thiazole-4-carboxylate ≥95%
Ethyl-2-(2,4-dichlorophenyl)thiazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₂H₉Cl₂NO₂S |
MDL Number:
MFCD09743715 CAS Number: 1185155-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...