2-(3,5-Dichlorophenyl)thiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

072059-1G 072059-5G
102522-880EA 269.23 USD
102522-880 102522-882
2-(3,5-Dichlorophenyl)thiazole-4-carboxylic acid ≥95%
2-(3,5-Dichlorophenyl)thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₅Cl₂NO₂S
MDL Number: MFCD07376343
CAS Number: 1178420-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR