Ethyl-2-(3,5-dichlorophenyl)thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(3,5-dichlorophenyl)thiazole-4-carboxylate

072058-1G 072058-5G
102522-876EA 710.77 USD
102522-876 102522-878
Ethyl-2-(3,5-dichlorophenyl)thiazole-4-carboxylate ≥95%
Ethyl-2-(3,5-dichlorophenyl)thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₉Cl₂NO₂S
MW: 302.18 g/mol
MDL Number: MFCD09743714
CAS Number: 132089-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR