5-Methyl-1H-indazole ≥95%

Supplier: Matrix Scientific

072054-250MG 072054-1G 072054-5G
102522-858EA 355.38 USD
102522-858 102522-860 102522-862
5-Methyl-1H-indazole ≥95%
5-Methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₈N₂
MW: 132.1600 g/mol
MDL Number: MFCD01606235
CAS Number: 1776-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR