Ethyl-1H-indazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1H-indazole-3-carboxylate

072053-1G 072053-5G
102522-854EA 710.77 USD
102522-854 102522-856
Ethyl-1H-indazole-3-carboxylate ≥95%
Ethyl 1H-indazole-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
MDL Number: MFCD01138134
CAS Number: 4498-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR