5-Bromo-1,3-dihydrobenzoimidazol-2-one ≥95%

Supplier: Matrix Scientific

072052-1G 072052-5G 072052-10G
102522-848EA 427.69 USD
102522-848 102522-850 102522-852
5-Bromo-1,3-dihydrobenzoimidazol-2-one ≥95%
5-Bromo-1,3-dihydrobenzoimidazol-2-one

Hazard: Irritant

Formula: C₇H₅BrN₂O
MDL Number: MFCD01893032
CAS Number: 39513-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR