5-Fluoro-1,3-dihydrobenzoimidazol-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-1H-benzo[d]imidazol-2(3H)-one

072051-1G 072051-5G
102522-844EA 167.69 USD
102522-844 102522-846
5-Fluoro-1,3-dihydrobenzoimidazol-2-one ≥95%
5-Fluoro-1,3-dihydrobenzoimidazol-2-one

Hazard: Irritant

Formula: C₇H₅FN₂O
MW: 152.13 g/mol
MDL Number: MFCD05663528
CAS Number: 1544-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR