4-(4-Methylpiperazin-1-ylmethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-((4-Methylpiperazin-1-yl)methyl)benzoic acid

072050-1G 072050-5G 072050-10G
102522-838EA 533.85 USD
102522-838 102522-840 102522-842
4-(4-Methylpiperazin-1-ylmethyl)benzoic acid ≥95%
4-(4-Methylpiperazin-1-ylmethyl)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MW: 234.3 g/mol
MDL Number: MFCD01632530
CAS Number: 106261-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR