[4-(5-Bromopyrimidin-2-yloxy)phenyl]methanol ≥95%

Supplier: Matrix Scientific

072049-1G 072049-5G 072049-10G
102522-832EA 355.38 USD
102522-832 102522-834 102522-836
[4-(5-Bromopyrimidin-2-yloxy)phenyl]methanol ≥95%
[4-(5-Bromopyrimidin-2-yloxy)phenyl]methanol

Hazard: Irritant

Formula: C₁₁H₉BrN₂O₂
MDL Number: MFCD09743713
CAS Number: 1189734-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR