1-Boc-4-(4-chloropyrimidin-2-yl)piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(4-chloropyrimidin-2-yl)piperazine-1-carboxylate

072048-1G 072048-5G
102522-828EA 463.08 USD
102522-828 102522-830
1-Boc-4-(4-chloropyrimidin-2-yl)piperazine ≥95%
1-Boc-4-(4-chloropyrimidin-2-yl)piperazine

Hazard: Irritant

Formula: C₁₃H₁₉ClN₄O₂
MW: 298.77 g/mol
MDL Number: MFCD09743711
CAS Number: 479691-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR