N-Boc-4'-methyl[1,4']bipiperidinyl ≥95%

Supplier: Matrix Scientific

072047-1G 072047-5G
102522-824EA 640 USD
102522-824 102522-826
N-Boc-4'-methyl[1,4']bipiperidinyl ≥95%
N-Boc-4'-methyl[1,4']bipiperidinyl

Hazard: Irritant

Formula: C₁₆H₃₀N₂O₂
MW: 282.43 g/mol
MDL Number: MFCD09743710
CAS Number: 864369-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR