2-(4-Nitrophenoxy)pyrimidine ≥95%

Supplier: Matrix Scientific

072041-1G 072041-5G 072041-10G
102522-788EA 355.38 USD
102522-788 102522-790 102522-792
2-(4-Nitrophenoxy)pyrimidine ≥95%
2-(4-Nitrophenoxy)pyrimidine

Hazard: Irritant

Formula: C₁₀H₇N₃O₃
MDL Number: MFCD00847151
CAS Number: 181801-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR