Methyl-4-(5-bromopyrimidin-2-yloxy)benzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-((5-bromopyrimidin-2-yl)oxy)benzoate
072039-1G
072039-5G
072039-10G
102522-776EA
355.38
USD
102522-776
102522-778
102522-780
Methyl-4-(5-bromopyrimidin-2-yloxy)benzoate ≥95%
Methyl-4-(5-bromopyrimidin-2-yloxy)benzoate
Hazard: Irritant
Formula:
C₁₂H₉BrN₂O₃ MW: 309.13 g/mol |
MDL Number:
MFCD09743706 CAS Number: 926304-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...