5-Bromo-2-(4-nitrophenoxy)pyrimidine ≥95%

Supplier: Matrix Scientific

072038-1G 072038-5G 072038-10G
102522-770EA 355.38 USD
102522-770 102522-772 102522-774
5-Bromo-2-(4-nitrophenoxy)pyrimidine ≥95%
5-Bromo-2-(4-nitrophenoxy)pyrimidine

Hazard: Irritant

Formula: C₁₀H₆BrN₃O₃
MDL Number: MFCD09743705
CAS Number: 1185158-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR