Ethyl-1-methyl-4-nitroimidazole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-methyl-4-nitro-1H-imidazole-2-carboxylate

072034-1G 072034-5G
102522-748EA 284.62 USD
102522-748 102522-750
Ethyl-1-methyl-4-nitroimidazole-2-carboxylate ≥95%
Ethyl-1-methyl-4-nitroimidazole-2-carboxylate

Hazard: Irritant

Formula: C₇H₉N₃O₄
MW: 199.17 g/mol
MDL Number: MFCD03789110
CAS Number: 109012-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR