1-Boc-4-methyl-4-morpholin-4-ylpiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-methyl-4-morpholinopiperidine-1-carboxylate

072033-1G 072033-5G
102522-744EA 640 USD
102522-744 102522-746
1-Boc-4-methyl-4-morpholin-4-ylpiperidine ≥95%
1-Boc-4-methyl-4-morpholin-4-ylpiperidine

Hazard: Irritant

Formula: C₁₅H₂₈N₂O₃
MW: 284.4 g/mol
MDL Number: MFCD09743700
CAS Number: 864369-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR