4-(Bromomethyl)benzenesulfonamide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromomethylbenzenesulphonamide

072032-1G 072032-5G
102522-740EA 612.31 USD
102522-740 102522-742
4-(Bromomethyl)benzenesulfonamide ≥95%
4-(Bromomethyl)benzenesulphonamide

Hazard: Irritant

Formula: C₇H₈BrNO₂S
MW: 250.1100 g/mol
MDL Number: MFCD08752696
CAS Number: 40724-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR