1-Boc-2-formylpyrrolidine ≥95%

Supplier: Matrix Scientific

072029-1G 072029-5G
102522-720EA 60 USD
102522-720 102522-722
1-Boc-2-formylpyrrolidine ≥95%
1-Boc-2-formylpyrrolidine

Hazard: Irritant/Store in Freezer

Formula: C₁₀H₁₇NO₃
MDL Number: MFCD03790896
CAS Number: 117625-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR