2-Amino-5-bromo-4-methoxypyrimidine ≥95%

Supplier: Matrix Scientific

072028-1G 072028-5G 072028-10G
102522-714EA 320 USD
102522-714 102522-716 102522-718
2-Amino-5-bromo-4-methoxypyrimidine ≥95%
2-Amino-5-bromo-4-methoxypyrimidine

Hazard: Irritant

Formula: C₅H₆BrN₃O
MDL Number: MFCD08460376
CAS Number: 36082-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR