(2-Chlorothiazol-5-yl)methylamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (2-Chlorothiazol-5-yl)methanamine hydrochloride

072026-1G 072026-5G 072026-10G
102522-702EA 107.69 USD
102522-702 102522-704 102522-706
(2-Chlorothiazol-5-yl)methylamine hydrochloride ≥95%
(2-Chlorothiazol-5-yl)methylamine hydrochloride

Hazard: Irritant

Formula: C₄H₆Cl₂N₂S
MW: 185.08 g/mol
MDL Number: MFCD09039253
CAS Number: 153471-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR