1-(4-Chloropyrimidin-2-yl)-4-piperidinol ≥95%

Supplier: Matrix Scientific

072025-1G 072025-5G
102522-698EA 355.38 USD
102522-698 102522-700
1-(4-Chloropyrimidin-2-yl)-4-piperidinol ≥95%
1-(4-Chloropyrimidin-2-yl)-4-piperidinol

Hazard: Irritant

Formula: C₉H₁₂ClN₃O
MDL Number: MFCD08752663
CAS Number: 916791-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR