5-Bromo-2-(pyrrolidin-3-yloxy)pyrimidine ≥95%

Supplier: Matrix Scientific

072024-1G 072024-5G 072024-10G
102522-692EA 533.85 USD
102522-692 102522-694 102522-696
5-Bromo-2-(pyrrolidin-3-yloxy)pyrimidine ≥95%
5-Bromo-2-(pyrrolidin-3-yloxy)pyrimidine

Hazard: Irritant

Formula: C₈H₁₀BrN₃O
MDL Number: MFCD08460370
CAS Number: 914347-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR