1-Boc-3-(5-bromopyrimidin-2-yloxy)pyrrolidine ≥95%

Supplier: Matrix Scientific

072023-1G 072023-5G 072023-10G
102522-686EA 284.62 USD
102522-686 102522-688 102522-690
1-Boc-3-(5-bromopyrimidin-2-yloxy)pyrrolidine ≥95%
1-Boc-3-(5-bromopyrimidin-2-yloxy)pyrrolidine

Hazard: Irritant

Formula: C₁₃H₁₈BrN₃O₃
MDL Number: MFCD08460369
CAS Number: 914347-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR