1-Boc-3-(5-bromopyrimidin-2-yloxy)piperidine ≥95%
Supplier: Matrix Scientific
072022-1G
072022-5G
072022-10G
102522-680EA
284.62
USD
102522-680
102522-682
102522-684
1-Boc-3-(5-bromopyrimidin-2-yloxy)piperidine ≥95%
1-Boc-3-(5-bromopyrimidin-2-yloxy)piperidine
Hazard: Irritant
Formula:
C₁₄H₂₀BrN₃O₃ |
MDL Number:
MFCD08460368 CAS Number: 914347-76-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...