5-Bromo-2-(piperidin-4-yloxy)pyrimidine ≥95%

Supplier: Matrix Scientific

072021-1G 072021-5G 072021-10G
102522-674EA 533.85 USD
102522-674 102522-676 102522-678
5-Bromo-2-(piperidin-4-yloxy)pyrimidine ≥95%
5-Bromo-2-(piperidin-4-yloxy)pyrimidine

Hazard: Irritant

Formula: C₉H₁₂BrN₃O
MDL Number: MFCD08460365
CAS Number: 792180-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR