5-Bromo-4-hydroxy-2-methylpyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-methylpyrimidin-4-ol

072016-1G 072016-5G
102522-648EA 144.62 USD
102522-648 102522-650
5-Bromo-4-hydroxy-2-methylpyrimidine ≥95%
5-Bromo-4-hydroxy-2-methylpyrimidine

Hazard: Irritant

Formula: C₅H₅BrN₂O
MW: 189.02 g/mol
MDL Number: MFCD08460358
CAS Number: 1676-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR