1-Boc-4-methyl-4-piperazin-1-ylpiperidine ≥95%
Supplier: Matrix Scientific
072015-250MG
072015-1G
072015-5G
102522-642EA
355.38
USD
102522-642
102522-644
102522-646
1-Boc-4-methyl-4-piperazin-1-ylpiperidine ≥95%
1-Boc-4-methyl-4-piperazin-1-ylpiperidine
Hazard: Irritant
Formula:
C₁₅H₂₉N₃O₂ MW: 283.42 g/mol |
MDL Number:
MFCD09743693 CAS Number: 1185064-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...