4'-Methyl[1,4']bipiperidinyl dihydrochloride ≥95%

Supplier: Matrix Scientific

072013-1G 072013-5G
102522-634EA 630.77 USD
102522-634 102522-636
4'-Methyl[1,4']bipiperidinyl dihydrochloride ≥95%
4'-Methyl[1,4']bipiperidinyl dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MW: 255.23 g/mol
MDL Number: MFCD09743691
CAS Number: 3543-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR