4-Amino-1-methylpiperidine ≥95%

Supplier: Matrix Scientific

072008-5G 072008-10G 072008-100G
102522-604EA 72.31 USD
102522-604 102522-606 102522-608
4-Amino-1-methylpiperidine ≥95%
4-Amino-1-methylpiperidine

Hazard: Irritant

Formula: C₆H₁₄N₂
MDL Number: MFCD01570549
CAS Number: 41838-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR