5-Methoxy-1H-indazole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

072005-1G 072005-5G 072005-10G
102522-586EA 273.85 USD
102522-586 102522-588 102522-590
5-Methoxy-1H-indazole-3-carbaldehyde ≥95%
5-Methoxy-1H-indazole-3-carbaldehyde

Hazard: Irritant

Formula: C₉H₈N₂O₂
MDL Number: MFCD06738288
CAS Number: 169789-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR