2-Amino-5-bromo-3-methoxypyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-3-methoxypyrazin-2-amine

072003-1G 072003-5G 072003-10G
102522-574EA 96.92 USD
102522-574 102522-576 102522-578
2-Amino-5-bromo-3-methoxypyrazine ≥95%
2-Amino-5-bromo-3-methoxypyrazine

Hazard: Irritant

Formula: C₅H₆BrN₃O
MW: 204.03 g/mol
MDL Number: MFCD08275726
CAS Number: 5900-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR