3-Bromo-7-hydroxy-4-methylchromen-2-one ≥95%

Supplier: Matrix Scientific

072002-1G 072002-5G 072002-10G
102522-568EA 284.62 USD
102522-568 102522-570 102522-572
3-Bromo-7-hydroxy-4-methylchromen-2-one ≥95%
3-Bromo-7-hydroxy-4-methylchromen-2-one

Hazard: Irritant

Formula: C₁₀H₇BrO₃
MDL Number: MFCD00510041
CAS Number: 55977-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR