4-(2-Fluoro-4-nitrophenoxy)piperidine hydrochloride ≥95%

Supplier: Matrix Scientific

072001-1G 072001-5G 072001-10G
102522-562EA 640 USD
102522-562 102522-564 102522-566
4-(2-Fluoro-4-nitrophenoxy)piperidine hydrochloride ≥95%
4-(2-Fluoro-4-nitrophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClFN₂O₃
MW: 276.69 g/mol
MDL Number: MFCD08448693
CAS Number: 1189653-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR