2-Iodo-4-methoxypyrimidine ≥95%

Supplier: Matrix Scientific

071999-1G 071999-5G 071999-10G
102522-550EA 160 USD
102522-550 102522-552 102522-554
2-Iodo-4-methoxypyrimidine ≥95%
2-Iodo-4-methoxypyrimidine

Hazard: Irritant

Formula: C₅H₅IN₂O
MDL Number: MFCD08275719
CAS Number: 262353-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR