Ethyl-4-hydroxyquinoline-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxyquinoline-3-carboxylic acid ethyl ester, Ethyl 4-hydroxyquinoline-3-carboxylate

071995-1G 071995-5G 071995-10G
102522-526EA 143.08 USD
102522-526 102522-528 102522-530
Ethyl-4-hydroxyquinoline-3-carboxylate ≥95%
Ethyl 4-hydroxyquinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MW: 217.2206 g/mol
MDL Number: MFCD00173406
CAS Number: 26892-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR