1-Boc-3-(3-methoxypropylamino)piperidine ≥95%

Supplier: Matrix Scientific

071994-1G 071994-5G
102522-522EA 640 USD
102522-522 102522-524
1-Boc-3-(3-methoxypropylamino)piperidine ≥95%
1-Boc-3-(3-methoxypropylamino)piperidine

Hazard: Irritant

Formula: C₁₄H₂₈N₂O₃
MDL Number: MFCD06656638
CAS Number: 887588-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR