2,4-Diaminopyrimidine-5-carbonitrile ≥95%

Supplier: Matrix Scientific

071993-1G 071993-5G 071993-10G
102522-516EA 355.38 USD
102522-516 102522-518 102522-520
2,4-Diaminopyrimidine-5-carbonitrile ≥95%
2,4-Diaminopyrimidine-5-carbonitrile

Hazard: Irritant

Formula: C₅H₅N₅
MDL Number: MFCD00085600
CAS Number: 16462-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR