5-Bromo-2-(dimethylamino)pyrimidine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-N,N-dimethylpyrimidin-2-amine
071991-1G
071991-5G
071991-10G
102522-506EA
161.54
USD
102522-506
102522-508
102522-510
5-Bromo-2-(dimethylamino)pyrimidine ≥95%
5-Bromo-2-(dimethylamino)pyrimidine
Hazard: Irritant
Formula:
C₆H₈BrN₃ MW: 202.0500 g/mol |
MDL Number:
MFCD07368228 CAS Number: 38696-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...