5-Bromo-2-(4-methylpiperazin-1-yl)pyrimidine ≥95%

Supplier: Matrix Scientific

071990-1G 071990-5G 071990-10G
102522-500EA 161.54 USD
102522-500 102522-502 102522-504
5-Bromo-2-(4-methylpiperazin-1-yl)pyrimidine ≥95%
5-Bromo-2-(4-methylpiperazin-1-yl)pyrimidine

Hazard: Irritant

Formula: C₉H₁₃BrN₄
MDL Number: MFCD03646018
CAS Number: 141302-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR