3-Amino-6-bromopyrazine-2-carbonitrile ≥95%

Supplier: Matrix Scientific

071986-1G 071986-5G
102522-476EA 90.77 USD
102522-476 102522-478
3-Amino-6-bromopyrazine-2-carbonitrile ≥95%
3-Amino-6-bromopyrazine-2-carbonitrile

Hazard: Irritant

Formula: C₅H₃BrN₄
MDL Number: MFCD06658300
CAS Number: 17231-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR