1-Benzyl-1H-indazol-5-ylamine ≥95%

Supplier: Matrix Scientific

071985-1G 071985-5G
102522-472EA 107.69 USD
102522-472 102522-474
1-Benzyl-1H-indazol-5-ylamine ≥95%
1-Benzyl-1H-indazol-5-ylamine

Hazard: Irritant

Formula: C₁₄H₁₃N₃
MW: 223.28 g/mol
MDL Number: MFCD01719807
CAS Number: 23856-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR