1-Benzyl-5-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

071984-1G 071984-5G 071984-10G
102522-466EA 427.69 USD
102522-466 102522-468 102522-470
1-Benzyl-5-nitro-1H-indazole ≥95%
1-Benzyl-5-nitro-1H-indazole

Hazard: Irritant

Formula: C₁₄H₁₁N₃O₂
MDL Number: MFCD08275706
CAS Number: 23856-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR