N-Boc-2-amino-4-formylthiazole ≥95%

Supplier: Matrix Scientific

Synonyms: N-Boc-2-Amino-4-formylthiazole

071983-1G 071983-5G 071983-10G
102522-460EA 533.85 USD
102522-460 102522-462 102522-464
N-Boc-2-amino-4-formylthiazole ≥95%
N-Boc-2-amino-4-formylthiazole

Hazard: Irritant

Formula: C₉H₁₂N₂O₃S
MW: 228.2682 g/mol
MDL Number: MFCD08275704
CAS Number: 494769-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR